Learn More
QuidelOrtho VITROS™ Calibrators: Testosterone
Description
VITROS™ MicroWell Calibrator designed for use with the Testosterone assay.
- Pack size: 100 wells
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | Testosterone |
| For Use With (Equipment) | VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system |
| Sample Type | Human Serum and Plasma (EDTA or Heparin) |
| Type | Testosterone Calibrator |
| Includes | 2mL VITROS™ Testosterone calibrators 1, 2 and 3 (Testosterone in Human Serum with antimicrobial agent), lot calibration card, protocol card, 24 calibrator bar code labels (8 for each calibrator) |
| Intended Use | Quantitative measurement of Testosterone |
| Level | 3 |
| Quantity | 1 Set/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.