Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho VITROS™ Calibrators: Total B-HCG II

Catalog No. OCD6802221
Change view
Click to view available options
Quantity:
3 Sets/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OCD6802221 3 Sets/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OCD6802221 Supplier QuidelOrtho Supplier No. 6802221

Specifications

Content And Storage 2°C to 8°C
For Use With (Equipment) VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system
Sample Type Human Serum and Plasma (Heparin and EDTA)
Type Total B-HCG II Calibrator
Includes VITROS™ Total beta-hCG II calibrators 1, 2 and 3, (freeze-dried, recombinant HCG in Human Plasma with antimicrobial agent, reconstitution volume 1mL), lot calibration card, protocol card, 24 calibrator bar code labels (8 for each calibra
Intended Use Quantitative measurement of Human Chorionic Gonadotropin (hCG) and its beta-subunit
Level 3
Lyophilized Yes
Quantity 3 Sets/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.