Learn More
QuidelOrtho VITROS™ Calibrators: Total T4
Description
The VITROS™ Total Thyroxine (tT4) Calibrator is intended for use in the calibration of the VITROS Immunodiagnostic Systems for the quantitative measurement of total thyroxine in human serum and plasma.
- Type: Calibrator
- Application: Quantitative measurement of total thyroxine
- Compatibility: VITROS Immunodiagnostic Systems
Specifications
Specifications
| Detectable Analytes | Total Thyroxine |
| For Use With (Equipment) | VITROS™ ECi immunodiagnostic system |
| Type | Total T4 Calibrator |
| Level | 3 |
| Quantity | 1 Set/Pk. |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.