Learn More
QuidelOrtho VITROS™ Controls: Anti-HBc IgM
Catalog No. OCD6801427
Change view
Click to view available options
Quantity:
0.5 mL
Catalog No. Quantity
OCD6801427 0.5 mL
1 options
Catalog No. OCD6801427 Supplier QuidelOrtho Supplier No. 6801427
Specifications
For Use With (Equipment) VITROS™ ECi immunodiagnostic system
Type Anti-HBc IgM Control
Intended Use Qualitative detection of IgM Antibody to Hepatitis B core Antigen (Anti-HBc IgM)
Quantity 0.5 mL