Learn More
QuidelOrtho VITROS™ Controls: Anti-HBc TOTAL
Catalog No. OCD6801430
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
OCD6801430 1 mL
1 options
Catalog No. OCD6801430 Supplier QuidelOrtho Supplier No. 6801430
Specifications
For Use With (Equipment) VITROS™ ECi immunodiagnostic system
Type Anti-HBc Control
Intended Use Qualitative detection of total Antibody (IgG and IgM) to Hepatitis B core Antigen (total Anti-HBc)
Quantity 1 mL