Learn More
QuidelOrtho VITROS™ Controls: Anti-HBs
Catalog No. OCD6802093
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
OCD6802093 2 mL
1 options
Catalog No. OCD6802093 Supplier QuidelOrtho Supplier No. 6802093
Specifications
For Use With (Equipment) VITROS™ ECi/ECiQ immunodiagnostic system
Type Anti-HBs Control
Intended Use Determination of total Antibody to Hepatitis B surface Antigen (Anti-HBs)
Quantity 2 mL