Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho VITROS™ ECiQ Immunodiagnostic System

Catalog No. 23057103
Change view
Click to view available options
Sample Size:
60 samples in universal sample trays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Size
23-057-103 60 samples in universal sample trays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-057-103 Supplier QuidelOrtho Supplier No. 1922814
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Depth (English) 29 in.
Product Type Immunodiagnostic System
Volume (Metric) Sample 10 to 80 μL
Depth (Metric) 73.6 cm
Frequency 47/63 Hz
Operating Temperature Range (English) 59°F to 86°F
Operating Temperature Range (Metric) 15°C to 30°C
Calibration Time 24 Days
Holds 10, 7, 5mL collection tubes , 1.5mL micro collection containers, Vitros™ microsample cup and 0.5 and 2mL cups
Electrical Requirements 120V/60 Hz, Single Phase 12.0 amp dedicated line
Includes Color coded graphical user interface, ergonomic flat, low-glare, LCD, touchscreen monitor, keyboard platform and support arm, flexible, customized positioning, on board documentation and help
Height (English) 51.25 in.
Height (Metric) 130.2 cm
Humidity 15 to 75%
Sample Size 60 samples in universal sample trays
Sample Type Serum, Plasma, Urine, Amniotic Fluid, Whole Blood
Voltage 120 V
Weight (English) 807 lb.
Weight (Metric) 366 kg
Width (English) 44 in.
Width (Metric) 111.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.