Learn More
QuidelOrtho VITROS™ Range Verifier: Estradiol
Catalog No. OCD6800380
Change view
Click to view available options
Quantity:
2 Pack
Catalog No. Quantity
OCD6800380 2 Pack
1 options
Catalog No. OCD6800380 Supplier QuidelOrtho Supplier No. 6800380
Add to Cart
Add to Cart
Verifies calibration range of VITROS immunodiagnostic systems
Specifications
Detectable Analytes Estradiol
Type Vitros™ Estradiol Verifier
Quantity 2 Pack