Learn More
QuidelOrtho VITROS™ Range Verifier: Follicle Stimulating Hormone (FSH)
Catalog No. OCD6800373
Change view
Click to view available options
Quantity:
2 Pack
Catalog No. Quantity
OCD6800373 2 Pack
1 options
Catalog No. OCD6800373 Supplier QuidelOrtho Supplier No. 6800373
Add to Cart
Add to Cart
Verifies calibration range of VITROS immunodiagnostic systems
Specifications
Detectable Analytes Follicle-Stimulating Hormone
Type Vitros™ Follicle Stimulating Hormone Verifier
Quantity 2 Pack