Learn More
QuidelOrtho VITROS™ Range Verifier: Free T4
Catalog No. OCD6800369
Change view
Click to view available options
Quantity:
2 Pack
Catalog No. Quantity
OCD6800369 2 Pack
1 options
Catalog No. OCD6800369 Supplier QuidelOrtho Supplier No. 6800369
Add to Cart
Add to Cart
Verifies calibration range of VITROS immunodiagnostic systems
Specifications
Detectable Analytes Free Thyroxine
Type Vitros™ Free T4 Verifier
Quantity 2 Pack