Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho VITROS™ Red Cell Folate Reagent

Catalog No. OCD1107804
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OCD1107804 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OCD1107804 Supplier QuidelOrtho Supplier No. 1107804
Add to Cart
Add to Cart

Specifications

Detectable Analytes RBC Folate
Sample Type EDTA Plasma, Heparin Plasma, Serum
Type Red Cell Folate Reagent
For Use With (Application) Quantitative measurement of red cell folate
Includes 2 Bottles of Red Cell Folate reagent with 27mL antimicrobial agent, 2 Red Cell Folate Ascorbate Tablets (54mg Ascorbic Acid/tablet) individually foil-wrapped, protocol card for red cell folate (colored pale blue)
Quantity 50 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.