Learn More
QuidelOrtho VITROS™ Thyroid-stimulating hormone (TSH)
Catalog No. OCD1912997
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD1912997 100 Pack
1 options
Catalog No. OCD1912997 Supplier QuidelOrtho Supplier No. 1912997
Specifications
Detectable Analytes Thyroid Stimulating Hormone
Format Well
Type Vitros™ TSH Reagent
Includes Coated wells 100, conjugate reagent 6.2mL, Biotinylated Antibody reagent 9.1mL
Quantity 100 Pack
Sample Size 80 μL
Test Time 37 min.