Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho VITROS™ 250/350 Solutions

Catalog No. OCD8262487
Change view
Click to view available options
Quantity:
12 x 5 mL
Type:
7% BSA Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
OCD8262487 12 x 5 mL 7% BSA Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OCD8262487 Supplier QuidelOrtho Supplier No. 8262487
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Solutions for QuidelOrtho Clinical VITROS 250/350

For use with QuidelOrtho VITROS 250 systems, Cat. Nos. 22-046-351, -353

Specifications

For Use With (Equipment) QuidelOrtho VITROS system
Type 7% BSA Solution
Quantity 12 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.