Learn More
QuidelOrtho Anti-A1 Lectin
SDP
Catalog No. 23252971 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
Quantity:
1 x 3 mL
Catalog No. Quantity
23-252971 1 x 3 mL
1 options
Catalog No. 23-252971 Supplier QuidelOrtho Supplier No. 711830
Add to Cart
Add to Cart
Designed for use in direct agglutination tests for recognition of the A1 antigen on human red blood cells
  • Purified extract of Dolichos biflorus seeds
Specifications
Type Anti-A1 Lectin
For Use With (Application) For use in direct Agglutination Tests for recognition of the A1 Antigen on Human red Blood cells
Quantity 1 x 3 mL