Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho QuidelOrtho VISION™ Analyzer
SDP

Catalog No. 23287100 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
Voltage:
100/240 VAC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
23-287-100 100/240 VAC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-287-100 Supplier QuidelOrtho Supplier No. 6904577
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Automates in vitro Immunohematology testing of human blood utilizing QuidelOrtho ID-MTS™ Gel card technology. QuidelOrtho Clinical Diagnostics QuidelOrtho VISION Analyzer provides responsive safety checks and balances give blood bankers the ability to monitor every critical step in the automation process.

Description

  • Column Agglutination Technology using QuidelOrtho ID-MTS™ Gel card technology
  • Automates test processing functions including liquid pipetting, reagent handling, incubation, centrifugation, reaction grading and interpretation and data management requirements using QuidelOrtho ID-MTS™ Gel card technology and digital image processing
  • Provides pre-qualification check of the front and back of every QuidelOrtho ID-MTS™ Gel card
  • Standalone instrument or interfaced to the Laboratory Information System (LIS)
  • Monitors critical processing monitoring and provides for complete management of system maintenance and quality control
  • Up to 42 samples can be loaded (six sample racks holding seven tubes each)
  • Authorized personnel on an external computer and on the laboratory's network can review results
  • Can be connected either to a network printer or to a local printer

QuidelOrtho VISION Supply Drawer

120 ID-MTS™ Gel Cards (6 sleeves of 20)

Reagent Red Blood Cell Supply

3 mL - 33 vials

10 mL - 18 vials

Any combination of 3 racks can be used to accommodate Reagent Red Blood Cell (RRBC) testing needs. The 10 mL rack can accommodate reverse grouping and antibody detection (screen) RRBC. The 3 mL rack can accommodate reverse group, antibody detection and Antibody Identification RRBC.

Diluent

2 x 100 mL for MTS™ Diluent 2 and MTS™ Diluent 2 Plus

2 x 10 mL for other diluents

e-Connectivity Remote Connectivity

Provides the ability to connect your system to QuidelOrtho in a way that enables remote diagnostics as well as, monitor and review system configuration, data, and performance.

Communication Ports

I/O ports include:

  • 1 DB-9 serial port (RS-232 port for LIS support)
  • 1 RJ45 LAN port supports port speeds of automatic, 10/100 Mbps with half and full duplex.

5 V 2.0/V 1.1 USB ports are available for:

  • Printer
  • Handheld barcode reader
  • Other devices

Specifications

  • Supported Sample Types: Centrifuged whole blood, plasma and serum, packed red blood cells
  • Sample and Test Processing: Continuous, random, STAT access and batch
  • Operating Temperature: 18°C to 30°C (64.4°F to 86°F)
  • Site Relative Humidity: 15 to 85% RH (non-condensing)
  • Maximum Altitude: 2438 m (8000 ft.)
  • Heat Output: 3412 BTU/hr.
  • Heated Incubator: 12 QuidelOrtho ID-MTS™ Gel Cards
  • Room Temperature Incubator: 16 QuidelOrtho ID-MTS™ Gel Cards
  • Centrifuge: two centrifuges; 10 QuidelOrtho ID-MTS™ Gel Cards per centrifuge
  • Waste: 5.2 L, 80 QuidelOrtho ID-MTS™ Gel Cards
  • Power Requirements: one dedicated (3-wired single phase; line-neutral and single circuit) AC power line for connection to facility power.
  • Input Voltage: 100 to 240V AC 50/60Hz 1000 VA
  • Dimensions (D x W x H): 77 x 107.4 x 88.9 cm (30.3 x 42.3 x 35 in.)
  • Weight: 190kg (419 lb.)

Applications

ABO/Rh Typing, Antibody Screen, Antibody Identification, Crossmatch, Direct Antiglobulin, Antigen Testing, QC testing, Serial Dilutions for Titration Studies, Monoclonal Rh Phenotyping, Selected Cell Panels

Specifications

Type Hematology Analyzer
Sample Type Centrifuged Whole Blood, Plasma and Serum, Packed Red Blood Cells
For Use With (Application) Automates in vitro Immunohematology testing of human blood utilizing ID-MTS™ Gel Card technology.
Depth (English) 30.3 in.
Depth (Metric) 107.4 cm
Height (English) 35 in.
Height (Metric) 88.9 cm
Width (English) 42.3 in.
Width (Metric) 107.4 cm
Electrical Requirements One dedicated (3-wired single phase; line-neutral and single circuit) AC power line for connection to facility power
Frequency 50/60 Hz
Voltage 100/240 VAC
Weight (Metric) 190 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.