Learn More
StatLab Ortho Xylene
Description
- Used to remove alcohol from tissue during processing, dissolve and remove paraffin wax before staining, and clear tissues to make them transparent for microscopic evaluation
- 4 gal. per Case
Specifications
Specifications
| Product Type | Clearing Agent |
| Quantity | 4 x 1 gal. |
| Form | Solution |