Learn More
QuidelOrtho VITROS™ Anti-Hepatitis C virus (A-HCV)
Catalog No. OCD6801325
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD6801325 100 Pack
1 options
Catalog No. OCD6801325 Supplier QuidelOrtho Supplier No. 6801325
Specifications
Detectable Analytes Hepatitis C Virus IgG
Format Well
Type Vitros™ Anti-HCV Reagent
Includes Coated wells 100, assay reagent 18.2mL, conjugate reagent 20.6mL
Quantity 100 Pack