Learn More
QuidelOrtho VITROS™ B12
Catalog No. OCD1453489
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD1453489 100 Pack
1 options
Catalog No. OCD1453489 Supplier QuidelOrtho Supplier No. 1453489
Specifications
Detectable Analytes Vitamin B12
Type Vitros™ Vitamin B12 Reagent
Includes Pack 1 includes coated wells 100, Biotinylated intrinsic factor reagent 6.7mL, conjugate reagent 6.7mL and Pack 2 includes uncoated wells 100, sample treatment reagent 6.2mL
Quantity 100 Pack
Sample Size 30 μL
Test Time 57 min.