Learn More
QuidelOrtho VITROS™ Carcinoembryonic antigen (CEA)
Catalog No. OCD1920115
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD1920115 100 Pack
1 options
Catalog No. OCD1920115 Supplier QuidelOrtho Supplier No. 1920115
Specifications
Detectable Analytes Carcinoembryonic Antigen
Format Well
Type Vitros™ CEA Reagent
Includes Coated wells 100, Biotinylated Antibody reagent 9.7mL, conjugate reagent 9.7mL
Quantity 100 Pack
Sample Size 20 μL
Test Time 30 min.