Learn More
QuidelOrtho VITROS™ Carcinoma Antigen (CA) 125II
Catalog No. OCD6801757
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD6801757 100 Pack
1 options
Catalog No. OCD6801757 Supplier QuidelOrtho Supplier No. 6801757
Specifications
Detectable Analytes OC 125 Antigen
Format Well
Type Vitros™ CA 125 II™ Reagent
Includes Coated wells 100, conjugate reagent 9.4mL, Biotinylated Antibody reagent 9.4mL
Quantity 100 Pack
Sample Size 25 μL
Test Time 37 min.