Learn More
QuidelOrtho VITROS™ Carcinoma Antigen (CA) 15-3
Catalog No. OCD6801758
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD6801758 100 Pack
1 options
Catalog No. OCD6801758 Supplier QuidelOrtho Supplier No. 6801758
Specifications
Detectable Analytes DF3 Antigen
Format Well
Type Vitros™ CA 15-3 Reagent
Includes Coated wells 100, conjugate reagent 20.2mL, Biotinylated Antibody reagent 15.8mL
Quantity 100 Pack
Test Time 42 min.