Learn More
QuidelOrtho VITROS™ Creatine Kinase-MB (CKMB)
Specifications
Specifications
| Detectable Analytes | CK-MB |
| Format | Well |
| Type | Vitros™ CK-MB Reagent |
| Includes | Coated wells 100, conjugate reagent 8.4mL, Biotinylated Antibody reagent 8.4mL |
| Quantity | 100 Pack |
| Sample Size | 40 μL |
| Test Time | 8 min. |