Learn More
QuidelOrtho VITROS™ Estradiol
Catalog No. OCD8552630
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD8552630 100 Pack
1 options
This item is not returnable. View return policy
Catalog No. OCD8552630 Supplier QuidelOrtho Supplier No. 8552630
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Detectable Analytes Estradiol
Format Well
Type Vitros™ Estradiol Reagent
Includes Coated wells 100, conjugate reagent 13.3mL, Biotinylated Antibody reagent 8.4mL
Quantity 100 Pack
Sample Size 25 μL
Test Time 40 min.