Learn More
QuidelOrtho VITROS™ Ferritin
Catalog No. OCD8356636
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD8356636 100 Pack
1 options
Catalog No. OCD8356636 Supplier QuidelOrtho Supplier No. 8356636
Specifications
Detectable Analytes Ferritin
Format Well
Type Vitros™ Ferritin Reagent
Includes Coated wells 100, Biotinylated Antibody reagent 19.4mL, conjugate reagent 19.4mL
Quantity 100 Pack
Sample Size 15 μL
Test Time 42 min.