Learn More
QuidelOrtho VITROS™ Folate
Catalog No. OCD1513266
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD1513266 100 Pack
1 options
Catalog No. OCD1513266 Supplier QuidelOrtho Supplier No. 1513266
Specifications
Detectable Analytes Folate
Format Well
Type Vitros™ Folate Reagent
Includes Coated wells 100, Biotinylated Folate binding Protein reagent 7.3mL, conjugate reagent 7.3mL
Quantity 100 Pack
Sample Size 53 μL
Test Time 73 min.