Learn More
QuidelOrtho VITROS™ Follicle-stimulating hormone (FSH)
Catalog No. OCD1931922
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD1931922 100 Pack
1 options
Catalog No. OCD1931922 Supplier QuidelOrtho Supplier No. 1931922
Specifications
Detectable Analytes Follicle-Stimulating Hormone
Format Well
Type Vitros™ FSH Reagent
Includes Coated wells 100, conjugate reagent 6.2mL, Biotinylated Antibody reagent 13.3mL
Quantity 100 Pack
Sample Size 40 μL
Test Time 24 min.