Learn More
QuidelOrtho VITROS™ Free T3
Catalog No. OCD1315589
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD1315589 100 Pack
1 options
Catalog No. OCD1315589 Supplier QuidelOrtho Supplier No. 1315589
Specifications
Detectable Analytes Free Triiodothyronine
Format Well
Type Vitros™ Free T3 Reagent
Includes Coated wells 100, conjugate reagent 13.3mL
Quantity 100 Pack
Sample Size 25 μL
Test Time 24 min.