Learn More
QuidelOrtho VITROS™ Free T4
Catalog No. OCD1387000
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD1387000 100 Pack
1 options
Catalog No. OCD1387000 Supplier QuidelOrtho Supplier No. 1387000
Specifications
Detectable Analytes Free Thyroxine
Format Well
Type Vitros™ Free T4 Reagent
Includes Coated wells 100, conjugate reagent 13.3mL
Quantity 100 Pack
Sample Size 25 μL
Test Time 24 min.