Learn More
QuidelOrtho VITROS™ Hepatitis B e Antigen (HBE AG)
Catalog No. OCD6801819
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD6801819 100 Pack
1 options
Catalog No. OCD6801819 Supplier QuidelOrtho Supplier No. 6801819
Specifications
Detectable Analytes Hepatitis B Antigen
Format Well
Type Vitros™ HBeAg Reagent
Includes Coated wells 100, assay reagent 6.6mL, conjugate reagent 6.6mL
Quantity 100 Pack
Sample Size 80 μL
Test Time 37 min.