Learn More
QuidelOrtho VITROS™ Luteinizing hormone (LH)
Catalog No. OCD1350198
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD1350198 100 Pack
1 options
Catalog No. OCD1350198 Supplier QuidelOrtho Supplier No. 1350198
Specifications
Detectable Analytes Luteinizing Hormone
Format Well
Type Vitros™ LH Reagent
Includes Coated wells 100, conjugate reagent 11mL, Biotinylated Antibody reagent 8.4mL
Quantity 100 Pack
Sample Size 50 μL
Test Time 24 min.