Learn More
QuidelOrtho VITROS™ N-terminal pro-brain natriuretic peptide
Shop All Ortho-Clinical Diagnostics Products
Specifications
Specifications
| Detectable Analytes | N-terminal pro-Brain Natriuretic Peptide |
| Format | Well |
| Type | Vitros™ NT-proBNP Reagent |
| Includes | Coated wells 100, conjugate reagent 8.2mL, Biotinylated Antibody reagent 8.2mL |
| Quantity | 100 Pack |
| Sample Size | 40 μL |
| Test Time | 16 min. |