Learn More
QuidelOrtho VITROS™ Progesteron
Catalog No. OCD1938570
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD1938570 100 Pack
1 options
Catalog No. OCD1938570 Supplier QuidelOrtho Supplier No. 1938570
Specifications
Detectable Analytes Progesterone
Format Well
Type Vitros™ Progesterone Reagent
Includes Coated wells 100, conjugate reagent 10mL, Biotinylated Antibody reagent 10mL
Quantity 100 Pack
Sample Size 25 μL
Test Time 24 min.