Learn More
QuidelOrtho VITROS™ Prostate-specific antigen (PSA)
Catalog No. OCD6801756
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD6801756 100 Pack
1 options
Catalog No. OCD6801756 Supplier QuidelOrtho Supplier No. 6801756
Specifications
Detectable Analytes Prostate specific Antigen
Format Well
Type Vitros™ PSA Reagent
Includes Coated wells 100, conjugate reagent 8.7mL, Biotinylated Antibody reagent 8.7mL
Quantity 100 Pack
Test Time 37 min.