Learn More
QuidelOrtho VITROS™ Rubella IGG
Catalog No. OCD8536195
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD8536195 100 Pack
1 options
Catalog No. OCD8536195 Supplier QuidelOrtho Supplier No. 8536195
Specifications
Detectable Analytes Rubella IgG
Format Well
Type Vitros™ Rubella IgG Reagent
Includes Coated wells 100, assay reagent 19.4mL, conjugate reagent 20.6mL
Quantity 100 Pack
Sample Size 10 μL
Test Time 35 min.