Learn More
QuidelOrtho VITROS™ Testosterone
Catalog No. OCD1435205
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD1435205 100 Pack
1 options
Catalog No. OCD1435205 Supplier QuidelOrtho Supplier No. 1435205
Specifications
Detectable Analytes Testosterone
Format Well
Type Vitros™ Testosterone Reagent
Includes Coated wells 100, conjugate reagent 8.4mL, Biotinylated Antibody reagent 13.3mL
Quantity 100 Pack
Sample Size 25 μL
Test Time 32 min.