Learn More
QuidelOrtho VITROS™ Total antibodies to Hepatitis B core antigen
Catalog No. OCD6801428
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD6801428 100 Pack
1 options
Catalog No. OCD6801428 Supplier QuidelOrtho Supplier No. 6801428
Specifications
Detectable Analytes Hepatitis B Antibodies
Format Well
Type Vitros™ Anti-HBc Reagent
Includes Coated wells 100, assay reagent 14.6mL, conjugate reagent 20.6mL
Quantity 100 Pack
Test Time 55 min.