Learn More
QuidelOrtho VITROS™ Total B-HCG II
Catalog No. OCD6802220
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD6802220 100 Pack
1 options
Catalog No. OCD6802220 Supplier QuidelOrtho Supplier No. 6802220
Specifications
Detectable Analytes Human chorionic gonadotropin
Format Well
Type Vitros™ Total Beta-hCG II Reagent
Includes Coated wells 100, Biotinylated Antibody reagent 14.4mL, conjugate reagent 19.2mL
Quantity 100 Pack
Sample Size 40 μL
Test Time 24 min.