Learn More
Ortho-Clinical Diagnostics™ VITROS™ Troponin I ES
SDP
Catalog No. 23057284 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
23-057-284 100 Pack
1 options
This item is not returnable. View return policy
Catalog No. 23-057-284 Supplier Ortho-Clinical Diagnostics™ Supplier No. 6802301
This item is not returnable. View return policy
Specifications
Detectable Analytes Cardiac Troponin I
Format Well
Type Vitros™ Troponin I ES Reagent
Includes Coated wells 100, conjugate reagent 7mL, Biotinylated Antibody reagent 7mL
Quantity 100 Pack
Sample Size 80 μL
Test Time 18 min.