Learn More
QuidelOrtho VITROS™ Troponin I ES
Catalog No. OCD6802301
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
OCD6802301 100 Pack
1 options
Catalog No. OCD6802301 Supplier QuidelOrtho Supplier No. 6802301
Specifications
Detectable Analytes Troponin I
Format Well
Type Vitros™ Troponin I ES Reagent
Includes Coated wells 100, conjugate reagent 7mL, Biotinylated Antibody reagent 7mL
Quantity 100 Pack
Sample Size 80 μL
Test Time 18 min.