Learn More
Med Vet International Orthopedic Drill & Accessories - Drill Set, J0238MN, Each
Catalog No. 5000585842
Encompass_Preferred
Orthopedic Power Driver. A power driver with features making it the best on the market. The power source is a high quality cordless reversible driver drill. - M12 12V Cordless Lithium-Ion 3/8 in. Drill - Includes 2 M12 REDLITHIUM CP1.5 Ah Compact Battery - M12 Li-ion Battery Charger - 2-speed gear box delivers 0-400 RPM on setting 1 and 0-1,500 RPM on setting 2 - Clutch adjusting ring for matching the torque setting to the application - LED work light illuminates dark work areas - Ergonomic handle for optimal user comfort and control - Battery fuel gauge displays remaining run-time so there's no guessing when the battery needs to be charged - Battery charge time - 30 minutes - Weight - 2.5 lbs. - Includes J0238s Drill shroud - Includes J0238d1 Power drill extension with chuck and key.
Catalog No. 50-005-85842 Supplier Med Vet International Supplier No. JOR238MN
Orthopedic Power Driver. A power driver with features making it the best on the market. The power source is a high quality cordless reversible driver drill. - M12 12V Cordless Lithium-Ion 3/8 in. Drill - Includes 2 M12 REDLITHIUM CP1.5 Ah Compact Battery - M12 Li-ion Battery Charger - 2-speed gear box delivers 0-400 RPM on setting 1 and 0-1,500 RPM on setting 2 - Clutch adjusting ring for matching the torque setting to the application - LED work light illuminates dark work areas - Ergonomic handle for optimal user comfort and control - Battery fuel gauge displays remaining run-time so there's no guessing when the battery needs to be charged - Battery charge time - 30 minutes - Weight - 2.5 lbs. - Includes J0238s Drill shroud - Includes J0238d1 Power drill extension with chuck and key.