Learn More
SPEX CERTIPREP LLC 3P WS-Orthophosphate
Catalog No. NC9807360
Encompass_Preferred
(NSI by Zeptometrix) A 21 mL concentrate for determination of Orthophosphate. Formulated in the NELAC range of 0.5 - 5.5 mg/L. Each ampule produces 2 liters of sample.
Catalog No. NC9807360 Supplier SPEX CERTIPREP LLC 3P Supplier No. PEI141
(NSI by Zeptometrix) A 21 mL concentrate for determination of Orthophosphate. Formulated in the NELAC range of 0.5 - 5.5 mg/L. Each ampule produces 2 liters of sample.