Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Orvepitant maleate | 579475-24-4 | 100.0% | 100 MG
SDP

Catalog No. 5000231203
Encompass_Preferred

Orvepitant maleate is the maleate salt of orvepitant, a potent and selective neurokinin-1 (NK1) receptor antagonist supplied as a high-purity solid for research use. It is suitable for in vitro and in vivo pharmacology, biochemical assays, and analytical studies.

  • High purity (99.99%) suitable for research applications.
  • Solid, off-white to light yellow appearance.
  • Molecular weight 744.70; molecular formula C35H39F7N4O6.
  • Available in multiple sizes, including 100 mg.
  • Storage: powder at -20°C (3 years) or 4°C (2 years).

Catalog No. 50-002-31203 Supplier Medchemexpress LLC Supplier No. HY122347A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Orvepitant maleate is the maleate salt of orvepitant, a potent and selective neurokinin-1 (NK1) receptor antagonist supplied as a high-purity solid for research use. It is suitable for in vitro and in vivo pharmacology, biochemical assays, and analytical studies.

  • High purity (99.99%) suitable for research applications.
  • Solid, off-white to light yellow appearance.
  • Molecular weight 744.70; molecular formula C35H39F7N4O6.
  • Available in multiple sizes, including 100 mg.
  • Storage: powder at -20°C (3 years) or 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.