Learn More
Medchemexpress LLC Orvepitant maleate | 579475-24-4 | 100.0% | 50 MG
SDP
Catalog No. 5000232492
Encompass_Preferred
Orvepitant maleate is the maleate salt of a potent and selective neurokinin-1 (NK-1) receptor antagonist used in preclinical research. It is orally active, crosses the blood-brain barrier, and has been investigated for indications including depressive disorders and chronic refractory cough.
  • Potent, selective NK-1 receptor antagonist with reported high binding affinity.
  • Orally active and capable of crossing the blood-brain barrier.
  • Supplied at high analytical purity for research applications.
  • Available in small milligram quantities for preclinical studies.
  • For laboratory research use only, not for human or veterinary use.
Catalog No. 50-002-32492 Supplier Medchemexpress LLC Supplier No. HY122347A50MG
Orvepitant maleate is the maleate salt of a potent and selective neurokinin-1 (NK-1) receptor antagonist used in preclinical research. It is orally active, crosses the blood-brain barrier, and has been investigated for indications including depressive disorders and chronic refractory cough.
  • Potent, selective NK-1 receptor antagonist with reported high binding affinity.
  • Orally active and capable of crossing the blood-brain barrier.
  • Supplied at high analytical purity for research applications.
  • Available in small milligram quantities for preclinical studies.
  • For laboratory research use only, not for human or veterinary use.