Learn More
Medchemexpress LLC Orvepitant maleate | 579475-24-4 | 99.0% | 25 MG
SDP
Catalog No. 5000232650
Encompass_Preferred
Orvepitant maleate is the maleate salt of an NK-1 receptor antagonist used for research. It is supplied as a powder for in vitro and in vivo studies, with high affinity for human NK-1 (pKi 10.2), DMSO solubility, and recommended cold storage for stability.
  • Maleate salt of an NK-1 receptor antagonist.
  • High affinity for human NK-1 (pKi 10.2).
  • Powder form with DMSO solubility of 30 mg/mL.
  • In vivo vehicle examples achieve ≥5 mg/mL clear solutions.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years).
Catalog No. 50-002-32650 Supplier Medchemexpress LLC Supplier No. HY122347A25MG
Orvepitant maleate is the maleate salt of an NK-1 receptor antagonist used for research. It is supplied as a powder for in vitro and in vivo studies, with high affinity for human NK-1 (pKi 10.2), DMSO solubility, and recommended cold storage for stability.
  • Maleate salt of an NK-1 receptor antagonist.
  • High affinity for human NK-1 (pKi 10.2).
  • Powder form with DMSO solubility of 30 mg/mL.
  • In vivo vehicle examples achieve ≥5 mg/mL clear solutions.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years).