Learn More
Apexbio Technology LLC ORY-1001 1431326-61-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063568
Encompass_Preferred
ORY-1001 is a selective inhibitor targeting the lysine-specific histone demethylase KDM1A (also known as LSD1) an FAD-dependent amine oxidase enzyme responsible for the removal of histone methylation marks particularly dimethylation of histone H3 lysine 4 (H3K4me2) By inhibiting KDM1A ORY-1001 promotes accumulation of H3K4me2 at gene targets and induces cellular differentiation in acute myeloid leukemia (AML) cell models In vitro assays demonstrated dose- and time-dependent increases in H3K4me2 and expression of differentiation markers in treated AML cells ORY-1001 exhibited oral bioavailability in animal xenograft models reducing tumor growth at low daily doses This molecule serves as a valuable research tool in epigenetics oncology pharmacology and studies regarding AML differentiation pathways with ongoing evaluation in early-phase clinical trials
Catalog No. 50-000-63568 Supplier Apexbio Technology LLC Supplier No. B58825.1
ORY-1001 is a selective inhibitor targeting the lysine-specific histone demethylase KDM1A (also known as LSD1) an FAD-dependent amine oxidase enzyme responsible for the removal of histone methylation marks particularly dimethylation of histone H3 lysine 4 (H3K4me2) By inhibiting KDM1A ORY-1001 promotes accumulation of H3K4me2 at gene targets and induces cellular differentiation in acute myeloid leukemia (AML) cell models In vitro assays demonstrated dose- and time-dependent increases in H3K4me2 and expression of differentiation markers in treated AML cells ORY-1001 exhibited oral bioavailability in animal xenograft models reducing tumor growth at low daily doses This molecule serves as a valuable research tool in epigenetics oncology pharmacology and studies regarding AML differentiation pathways with ongoing evaluation in early-phase clinical trials