Learn More
Medchemexpress LLC γ-Oryzanol | 11042-64-1 | 99.66% | 500 MG
SDP
Catalog No. 5000410913
Encompass_Preferred
γ-Oryzanol is a potent DNA methyltransferases (DNMTs) inhibitor in the striatum of mice. It significantly inhibits the activities of DNMT1 and DNMT3a.
  • Potent DNA methyltransferases inhibitor.
  • Significantly inhibits DNMT1 activity (IC50=3.2 μM).
  • Significantly inhibits DNMT3a activity (IC50=22.3 μM).
Catalog No. 50-004-10913 Supplier Medchemexpress LLC Supplier No. HYB2194500MG
γ-Oryzanol is a potent DNA methyltransferases (DNMTs) inhibitor in the striatum of mice. It significantly inhibits the activities of DNMT1 and DNMT3a.
  • Potent DNA methyltransferases inhibitor.
  • Significantly inhibits DNMT1 activity (IC50=3.2 μM).
  • Significantly inhibits DNMT3a activity (IC50=22.3 μM).