Learn More
Medchemexpress LLC OS-2966 5mg | 5mg
SDP
Catalog No. 5000324898
Encompass_Preferred
OS-2966 is a humanized antibody expressed in CHO cells targeting Integrin b1/ITGB1/CD29 OS-2966 carries a huIgG1 type heavy chain and a hu type light chain with a predicted molecular weight (MW) of 145 5 kDa The isotype control for OS-2966 can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-24898 Supplier Medchemexpress LLC Supplier No. HYP9906435MG
OS-2966 is a humanized antibody expressed in CHO cells targeting Integrin b1/ITGB1/CD29 OS-2966 carries a huIgG1 type heavy chain and a hu type light chain with a predicted molecular weight (MW) of 145 5 kDa The isotype control for OS-2966 can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)