Learn More
Medchemexpress LLC OS-3-106 100mg | 1580000-17-4 | 450.60 | C25H30N4O2S | 100 MG
SDP
Catalog No. 5000228269
Encompass_Preferred
OS-3-106 is a potent, blood-brain barrier-penetrant, selective dopamine D3 receptor (D3R) agonist with high affinity (Ki = 0.2 nM). It is supplied as a research solid and is used in neuropharmacology and addiction-related studies.
  • Selective dopamine D3 receptor agonist with Ki = 0.2 nM.
  • Blood-brain barrier penetrant suitable for central nervous system studies.
  • Reported high purity (97.03%), appropriate for research use.
  • Available in multiple pack sizes including 100 MG.
  • Provided as a solid suitable for in vitro and in vivo experiments.
Catalog No. 50-002-28269 Supplier Medchemexpress LLC Supplier No. HY116820100MG
OS-3-106 is a potent, blood-brain barrier-penetrant, selective dopamine D3 receptor (D3R) agonist with high affinity (Ki = 0.2 nM). It is supplied as a research solid and is used in neuropharmacology and addiction-related studies.
  • Selective dopamine D3 receptor agonist with Ki = 0.2 nM.
  • Blood-brain barrier penetrant suitable for central nervous system studies.
  • Reported high purity (97.03%), appropriate for research use.
  • Available in multiple pack sizes including 100 MG.
  • Provided as a solid suitable for in vitro and in vivo experiments.