Learn More
Medchemexpress LLC OS-3-106 | 10MG
SDP
Catalog No. 5000227879
Encompass_Preferred
OS-3-106 is a potent BBB-penetrated and selective dopamine D3 receptor (D3R) agonist OS-3-106 binds with high affinity (Ki 0 2 nM) at the D3R OS-3-106 can be used for psychoactivator addiction research[1]
Catalog No. 50-002-27879 Supplier Medchemexpress LLC Supplier No. HY11682010MG
OS-3-106 is a potent BBB-penetrated and selective dopamine D3 receptor (D3R) agonist OS-3-106 binds with high affinity (Ki 0 2 nM) at the D3R OS-3-106 can be used for psychoactivator addiction research[1]