Learn More
Medchemexpress LLC OS-3-106 25mg | 1580000-17-4 | 450.60 g/mol | C25H30N4O2S | 25 MG
SDP
Catalog No. 5000227232
Encompass_Preferred
OS-3-106 is a potent, blood-brain barrier penetrant dopamine D3 receptor (D3R) agonist with high affinity (Ki = 0.2 nM) and approximately 115-fold selectivity over D2R. It is supplied as a purified research compound for in vitro and in vivo pharmacology studies.
  • High affinity D3R agonist (Ki = 0.2 nM).
  • Approximately 115-fold selectivity for D3R versus D2R.
  • Blood-brain barrier penetrant, suitable for CNS studies.
  • High purity (97.03%).
  • Supplied as a small-quantity research pack (25 mg).
Catalog No. 50-002-27232 Supplier Medchemexpress LLC Supplier No. HY11682025MG
OS-3-106 is a potent, blood-brain barrier penetrant dopamine D3 receptor (D3R) agonist with high affinity (Ki = 0.2 nM) and approximately 115-fold selectivity over D2R. It is supplied as a purified research compound for in vitro and in vivo pharmacology studies.
  • High affinity D3R agonist (Ki = 0.2 nM).
  • Approximately 115-fold selectivity for D3R versus D2R.
  • Blood-brain barrier penetrant, suitable for CNS studies.
  • High purity (97.03%).
  • Supplied as a small-quantity research pack (25 mg).